Phosphorus, Oil based standard solution, Specpure™ P 1000μg/g
Phosphorus, Oil based standard solution, Specpure™ P 1000μg/g
Thermo Scientific Chemicals
Phosphorus, Oil based standard solution, Specpure™ P 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014210.1850 g
Catalog number ALF014210.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS17715-00-3
IUPAC Name(prop-2-yn-1-yl)cyclohexane
Molecular FormulaC9H14
InChI KeyUARFKZSJGDQRLF-UHFFFAOYSA-N
SMILESC#CCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4560-1.4610 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%