Hypophosphorous Acid, 50% w/w Aqueous Solution, Thermo Scientific™
Hypophosphorous Acid, 50% w/w Aqueous Solution, Thermo Scientific™
Hypophosphorous Acid, 50% w/w Aqueous Solution, Thermo Scientific™
Hypophosphorous Acid, 50% w/w Aqueous Solution, Thermo Scientific™
Thermo Scientific Chemicals
Hypophosphorous Acid, 50% w/w Aqueous Solution, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALF014142.574 x 500 g
Catalog number ALF014142.57
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHypophosphorous acid
ColorColorless
Name Note50% w/w aq. soln.
CAS6303-21-5
Health Hazard 1Danger
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
  • Primarily used for electroless nickel plating.
  • Involved in the reduction of arenediazonium salts.
  • As an additive in Fischer esterification reactions.
  • Serves as a neutralizing agent, antioxidant, catalyst in polymerization and poly condensation, and wetting agent.
  • Used in the formulation of pharmaceuticals, discoloration of polymers, water treatment and retrieval of precious or non-ferrous metals.
  • Used as bleaching agents for plastics, synthetic fibers, decolorizing agent and for color stabilization during the manufacture of chemicals and several plastics
Incompatible with strong oxidizers, mercuric oxide, mercury(II) nitrate, metals and strong bases