Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
5 cm
Catalog number ALF013996.BU
Price (MYR)
3,475.00
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (MYR)
3,475.00
Add to cart
Specifications
Chemical Name or MaterialVanadium rod
Name Note6.35mm (0.25 in.) dia.
CAS7440-62-2
Recommended StorageAmbient temperatures
EINECS Number231-171-1