Titanium foil, 0.127mm (0.005in) thick, 99.99+% (metals basis), Thermo Scientific Chemicals
Titanium foil, 0.127mm (0.005in) thick, 99.99+% (metals basis), Thermo Scientific Chemicals
Titanium foil, 0.127mm (0.005in) thick, 99.99+% (metals basis), Thermo Scientific Chemicals
Titanium foil, 0.127mm (0.005in) thick, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium foil, 0.127mm (0.005in) thick, 99.99+% (metals basis), Thermo Scientific Chemicals
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF013976.FI
Price (MYR)
1,315.00
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
1,315.00
Add to cart
Specifications
Chemical Name or MaterialTitanium foil
Name Note0.127mm (0.005 in.) thick
CAS7440-32-6
Recommended StorageAmbient temperatures
EINECS Number231-142-3