Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL, Thermo Scientific Chemicals
Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL, Thermo Scientific Chemicals
Sr(NO3)2 in 5% HNO3
Have Questions?
Catalog NumberQuantity
ALF013874.AE100 mL
Catalog number ALF013874.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS111-82-0
IUPAC Namemethyl dodecanoate
Molecular FormulaC13H26O2
InChI KeyUQDUPQYQJKYHQI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4290 to 1.4340 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %