Phosphorus plasma standard solution, Thermo Scientific™, Specpure™
Have Questions?
Catalog Number
Quantity
ALF013862.AE
100 mL
Catalog number ALF013862.AE
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphorus plasma standard solution
Name NoteP 1000 μg/mL
Quantity100 mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement H314 Causes severe skin burns and eye damage.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.