Potassium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
For Pyrotechnics, Glass Manufacture, Tempering Steel
Have Questions?
Catalog NumberQuantity
ALF013443.22100 g
Catalog number ALF013443.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS87151-66-4
IUPAC Name3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Molecular FormulaC9H14O
InChI KeyDJJNAQALDCPMOU-UHFFFAOYSA-N
SMILESOCCCC1=CCC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Refractive Index1.5040-4.5090 @ 20?C
Appearance (Color)Clear, colorless
FormLiquid
Impurity content3-Phenylpropanol content: <3.0% by NMR
Pyrotechnics, glass manufacture, tempering steel, reagentPotassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.