Boric acid, 99.8%, Thermo Scientific Chemicals
Boric acid, 99.8%, Thermo Scientific Chemicals
Boric acid, 99.8%, Thermo Scientific Chemicals
Boric acid, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.8%, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantity
ALF012680.A75 kg
Catalog number ALF012680.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS16373-93-6
IUPAC Name(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H13N5O3
InChI KeyOLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILESNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Water=<8 % (K.F.)
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.