Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
CAS: 14363-15-6 | C10H16O5Zn | 281.61 g/mol
Have Questions?
Catalog NumberQuantity
ALF012527.30250 g
Catalog number ALF012527.30
Price (MYR)
805.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
805.00
Add to cart
Specifications
Chemical Name or MaterialZinc 2,4-pentanedionate monohydrate
CAS14363-15-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.