Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 12069-89-5 | CMo2 | 203.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF012192.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS25952-53-8
IUPAC Name({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular FormulaC8H18ClN3
InChI KeyFPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILESCl.CCN=C=NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.0%
Appearance (Color)White
Assay (Titration ex Chloride)≥98.0 to ≤102.0% (non-U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
View more
Molybdenum carbide is used in the production of solid molybdenum carbide grades of hard metal in the manufacture of mixed crystal. It used as coating material.