Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Chelating agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF011931.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS4870-65-9
IUPAC Name2-bromo-2-phenylacetic acid
Molecular FormulaC8H7BrO2
InChI KeyWAKFRZBXTKUFIW-UHFFFAOYNA-N
SMILESOC(=O)C(Br)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.