Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Chelating agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF011931.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS95-45-4
IUPAC Name(E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
Molecular FormulaC4H8N2O2
InChI KeyJGUQDUKBUKFFRO-GGWOSOGESA-N
SMILESC\C(=N/O)\C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Suitability(for Ni determination) Passes test
Infrared spectrumConforms
Gravimetry>=99.0 %
View more
Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.