SiliaPrepX™ SPE Cartridges - WAX
SiliaPrepX™ SPE Cartridges - WAX
Actual product may vary
Thermo Scientific Chemicals
SiliaPrepX™ SPE Cartridges - WAX
SPE Cartridges packed with a polymeric weak anion exchange (WAX) phase, for extraction of strong acidic compounds.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightVolume (Metric)Quantity
ALF011846.0960 mg1 mL100 Pk.
ALF011846.18200 mg3 mL50 Pk.
Catalog number ALF011846.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
60 mg
Volume (Metric):
1 mL
Quantity:
100 Pk.
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Adsorbent TypePolymer
Carbon Load0.86
Bed Weight60 mg
For Use With (Application)Extraction of strongly acidic compounds, highly stable in organic solvents
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size85 μm
Product LineSiliaPrepX
SorbentPolystyrene-divinylbenzene WAX
TypeSPE Cartridge
Volume (Metric)1 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeSpherical
Pore Size60 Å
Quantity100 Pk.
Surface Area (Metric)800 m2/g
Tube Size5.7 x 65.7 mm
Length (Metric)65.7 mm
Diameter (Metric) Inner5.7 mm
Unit Size