SiliaSep OT Flash Cartridges, Propylsulfonic Acid (SCX-2)
SiliaSep OT Flash Cartridges, Propylsulfonic Acid (SCX-2)
Actual product may vary
Thermo Scientific Chemicals
SiliaSep OT Flash Cartridges, Propylsulfonic Acid (SCX-2)
Open-top Flash Cartridges packed with Propylsulfonic Acid sorbent (SCX-2) for purification of weak basic molecules.
Have Questions?
Catalog NumberBed WeightDiameter (Metric) InnerLength (Metric)Volume (Metric)
ALF011827.1850 g38.2 mm170 mm150 mL
Catalog number ALF011827.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
50 g
Diameter (Metric) Inner:
38.2 mm
Length (Metric):
170 mm
Volume (Metric):
150 mL
SiliaSep OT Flash Cartridges are for use with vacuum manifolds and automated SPE equipments.
  • Also compatible with FlashMaster™ systems
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing for excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
  • Maximum operating pressure: 60 psi
Specifications
Adsorbent TypeSilica
Carbon Load5%
Column TypeOpen-top SPE Cartridge
Bed Weight50 g
For Use With (Application)Extraction of weak basic molecules, catch and release of amines
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size40 to 63 μm
Product LineSiliaSep OT
SorbentPropylsulfonic Acid (SCX-2)
TypeSPE Cartridge
Volume (Metric)150 mL
EndcappedYes
MaterialPolypropylene tube, Polyethylene frits
Particle ShapeIrregular
Pore Size60 Å
Quantity10/box
Surface Area (Metric)500 m2/g
Length (Metric)170 mm
Diameter (Metric) Inner38.2 mm
Unit Size