Palladium foil, 0.25mm (0.01in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium foil, 0.25mm (0.01in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium foil, 0.25mm (0.01in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium foil, 0.25mm (0.01in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium foil, 0.25mm (0.01in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number ALF011516.GH
Price (MYR)
44,417.00
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
44,417.00
Add to cart
Specifications
Chemical Name or MaterialPalladium foil
Name Note0.25mm (0.01 in.) thick
CAS7440-05-3
Recommended StorageAmbient temperatures
EINECS Number231-115-6