Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF011507.FF25 x 25 mm
Catalog number ALF011507.FF
Price (MYR)
12,543.00
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (MYR)
12,543.00
Add to cart
Specifications
Chemical Name or MaterialPlatinum
CAS7440-06-4
AppearanceSilvery white foil
Recommended StorageAmbient temperatures
EINECS Number231-116-1