Silver powder, APS 4-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, APS 4-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, APS 4-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, APS 4-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver powder, APS 4-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Form: S.A. 0.1-0.4m2/g
Have Questions?
Catalog NumberQuantity
ALF011402.1850 g
Catalog number ALF011402.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1317-39-1
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.