Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals
Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals
Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals
Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals
CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number ALF011370.GH
Price (MYR)
1,216.00
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
1,216.00
Add to cart
Specifications
Chemical Name or MaterialCadmium foil
Name Note0.25mm (0.01 in.) thick
CAS7440-43-9
Health Hazard 1H330-H341-H350-H361fd-H372-H500
Health Hazard 2GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.