n-Tetracontane, 97% min, Thermo Scientific Chemicals
n-Tetracontane, 97% min, Thermo Scientific Chemicals
n-Tetracontane, 97% min, Thermo Scientific Chemicals
n-Tetracontane, 97% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Tetracontane, 97% min, Thermo Scientific Chemicals
CAS: 4181-95-7 | C40H82 | 563.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF011274.042 g
Catalog number ALF011274.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS16115-80-3
IUPAC Namehydrogen 1,3-dimethyl 2-aminopropanedioate chloride
Molecular FormulaC5H10ClNO4
InChI KeyQWNDKNJSEWOEDM-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC(=O)C(N)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Color)Light yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).