Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
2 cm
Catalog number ALF011098.BS
Price (MYR)
5,405.00
Add to cart
Quantity:
10 cm
Request bulk or custom format
Price (MYR)
5,405.00
Add to cart
Specifications
Chemical Name or MaterialVanadium rod
Name Note12.7mm (0.5 in.) dia.
CAS7440-62-2
Recommended StorageAmbient temperatures
EINECS Number231-171-1