Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 mL
100 mL
Catalog number ALF011000.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS50-85-1
IUPAC Name2-hydroxy-4-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyNJESAXZANHETJV-UHFFFAOYSA-N
SMILESCC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point171°C to 179°C
Appearance (Color)White to grey-beige
Titration with NaOH>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, insecticides, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.