Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF010958.BW50 cm
Catalog number ALF010958.BW
Price (MYR)
1,622.00
Add to cart
Quantity:
50 cm
Request bulk or custom format
Price (MYR)
1,622.00
Add to cart
Specifications
Chemical Name or MaterialPlatinum
CAS7440-06-4
AppearanceSilvery white wire
Recommended StorageAmbient temperatures
EINECS Number231-116-1