Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF010956.BP2 cm
Catalog number ALF010956.BP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 cm
Request bulk or custom format
Chemical Identifiers
CAS13512-31-7
IUPAC Namemethyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC18H19NO5
InChI KeySLLMDHBKALJDBW-XISACWJONA-N
SMILESCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Supplier's CofA≥97.5%