Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Catalog NumberQuantity
ALF010755.1850 g
Catalog number ALF010755.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS706-14-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)>97.5%
FormLiquid
Refractive Index1.4470-1.4510 @ 20?C