Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 13778-31-9 | CuN2O6 | 187.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALF010699.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS69365-72-6
IUPAC Namegallium(3+) trinitrate
Molecular FormulaGaN3O9
InChI KeyCHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Total trace metal impurities=<0.0002 %
Appearance (Color)White
Gallium (Ga)18 to 22 %
It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.