Gold powder, Premion™ 99.99% (metals basis), Thermo Scientific Chemicals
Gold powder, Premion™ 99.99% (metals basis), Thermo Scientific Chemicals
Gold powder, Premion™ 99.99% (metals basis), Thermo Scientific Chemicals
Gold powder, Premion™ 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold powder, Premion™ 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF010612.031 g
Catalog number ALF010612.03
Price (MYR)
1,836.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,836.00
Add to cart
Specifications
Chemical Name or MaterialGold powder
ColorGolden
CAS7440-57-5
Recommended StorageAmbient temperatures
RTECS NumberMD5070000