Cobalt pieces, 99.9+% (metals basis), Thermo Scientific Chemicals
Cobalt pieces, 99.9+% (metals basis), Thermo Scientific Chemicals
Cobalt pieces, 99.9+% (metals basis), Thermo Scientific Chemicals
Cobalt pieces, 99.9+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt pieces, 99.9+% (metals basis), Thermo Scientific Chemicals
CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Catalog NumberQuantity
ALF010454.22100 g
Catalog number ALF010454.22
Price (MYR)
701.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
701.00
Add to cart
Specifications
Chemical Name or MaterialCobalt
ColorGray
CAS7440-48-4
Health Hazard 1H317-H334-H350
Health Hazard 3P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c