Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Catalog NumberQuantity
ALF010411.G925 m
Catalog number ALF010411.G9
Price (MYR)
4,829.00
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (MYR)
4,829.00
Add to cart
Specifications
Chemical Name or MaterialTungsten
CAS7440-33-7
AppearanceSilver Gray wire
Recommended StorageAmbient temperatures
EINECS Number231-143-9