Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Catalog NumberQuantity
ALF010350.G55 m
Catalog number ALF010350.G5
Price (MYR)
2,367.00
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (MYR)
2,367.00
Add to cart
Specifications
Chemical Name or MaterialTantalum wire
Name Note1.0mm (0.04 in.) dia., Annealed, Temper: soft
CAS7440-25-7
Recommended StorageAmbient temperatures
EINECS Number231-135-5