Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Form: Approx.6.7g/m
Have Questions?
Catalog NumberQuantity
ALF010264.G55 m
Catalog number ALF010264.G5
Price (MYR)
926.00
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (MYR)
926.00
Add to cart
Specifications
Chemical Name or MaterialNiobium
ColorSilver to Gray
CAS7440-03-1
AppearanceSilver gray wire