Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 x 500 mm
50 x 100 mm
100 x 200 mm
Catalog number ALF010259.HQ
Price (MYR)
2,900.00
Add to cart
Quantity:
200 x 500 mm
Request bulk or custom format
Price (MYR)
2,900.00
Add to cart
Specifications
Chemical Name or MaterialNiobium foil
Name Note0.25mm (0.01 in.) thick, Annealed
CAS7440-03-1
Recommended StorageAmbient temperatures
EINECS Number231-113-5