Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF010205.BY100 cm
Catalog number ALF010205.BY
Price (MYR)
937.00
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (MYR)
937.00
Add to cart
Specifications
Chemical Name or MaterialHafnium wire
Name Note1.0mm (0.04 in.) dia.
CAS7440-58-6
Recommended StorageAmbient temperatures
EINECS Number231-166-4