Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Catalog NumberQuantity
ALF010187.1850 g
Catalog number ALF010187.18
Price (MYR)
2,454.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
2,454.00
Add to cart
Specifications
Chemical Name or MaterialGallium ingot
ColorGray-Silver
CAS7440-55-3
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H290
May be corrosive to metals.