Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF010161.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored strands
Recommended StorageAmbient temperatures
EINECS Number231-159-6