Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF010159.A3
Price (MYR)
1,194.00
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
1,194.00
Add to cart
Specifications
Chemical Name or MaterialCopper shot
Name Note1 to 10mm (0.04 to 0.4 in.)
CAS7440-50-8
Recommended StorageAmbient temperatures
EINECS Number231-159-6