Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-38-2 | As | 74.92 g/mol
Have Questions?
Catalog NumberQuantity
ALF010101.1850 g
Catalog number ALF010101.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl99.7 to 100.3 % (On dried substance)
Heavy metals (as Pb)=<5 ppm
Appearance (Color)White
Appearance (Form)Powder
Sulfur compounds (as SO4)=<0.003 %