Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Catalog NumberQuantity
ALF010100.A11 kg
Catalog number ALF010100.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Residual water=<2 % (150°C)
Appearance (Form)Beads
Water adsorption>=20 % (50% rH, 1 bar, 25°C)
ColorOrange (dry) to Colorless (wet)