Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF010093.A7
Price (MYR)
1,759.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
1,759.00
Add to cart
Specifications
Chemical Name or MaterialAluminum granules
Name Note8 to 12mm (0.32 to 0.47 in.)
CAS7429-90-5
Recommended StorageAmbient temperatures
EINECS Number231-072-3