Aluminum lump, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum lump, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum lump, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum lump, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum lump, 99.5% (metals basis), Thermo Scientific Chemicals
Form: Approx. 17g/lump
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number ALF010092.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS99747-74-7
IUPAC Namenaphthalen-1-yl trifluoromethanesulfonate
Molecular FormulaC11H7F3O3S
InChI KeyWQWUQDVFRYMMCY-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)OC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%