Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF000501.FS25 x 150 mm
Catalog number ALF000501.FS
Price (MYR)
733.00
Add to cart
Quantity:
25 x 150 mm
Request bulk or custom format
Price (MYR)
733.00
Add to cart
Specifications
Chemical Name or MaterialSilver foil
Name Note0.075mm (0.003 in.) thick, Hard, Temper: as rolled
CAS7440-22-4
Recommended StorageAmbient temperatures
EINECS Number231-131-3