Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Iron powder, spherical, &lt;20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Iron powder, spherical, &lt;20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Iron powder, spherical, &lt;20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Iron powder, spherical, &lt;20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF000170.30250 g
Catalog number ALF000170.30
Price (MYR)
538.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
538.00
Add to cart
Specifications
Chemical Name or MaterialIron
CAS7439-89-6
Health Hazard 1H500-X
Health Hazard 3P235+P410-P280
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
RUO – Research Use Only