Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR610350010
Price (MYR)
2,074.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,074.00
Add to cart
Chemical Identifiers
CAS64-04-0
IUPAC Name2-phenylethan-1-amine
Molecular FormulaC8H11N
InChI KeyBHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILESNCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.5 %
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow