Locust bean gum
Thermo Scientific Chemicals
Locust bean gum
CAS: 9000-40-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR466672500
Price (MYR)
668.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
668.00
Add to cart
Chemical Identifiers
CAS74-88-4
IUPAC Nameiodomethane
Molecular FormulaCH3I
InChI KeyINQOMBQAUSQDDS-UHFFFAOYSA-N
SMILESCI
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Non-volatile matter=<0.01 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.5290 to 1.5310 (20°C, 589 nm)