Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number ACR466090050
Price (MYR)
307.85
EA
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
307.85
EA
Add to cart
Chemical Identifiers
CAS776-74-9
IUPAC Name[bromo(phenyl)methyl]benzene
Molecular FormulaC13H11Br
InChI KeyOQROAIRCEOBYJA-UHFFFAOYSA-N
SMILESBrC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
GC>=88 %
Appearance (Form)Low melting crystalline mass or solid
Infrared spectrumConforms
Melting point33°C to 42°C