Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Talc, specified acc.to the requirements of Ph.Eur, USP, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number ACR466090010
Price (MYR)
102.81
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
102.81
EA
Add to cart
Specifications
Chemical Name or MaterialTalc
Melting Point800.0°C
CAS14807-96-6
Quantity1 kg
EINECS Number238-877-9