Toluene-d8, for NMR, 99.8 atom % D
Toluene-d8, for NMR, 99.8 atom % D
Thermo Scientific Chemicals
Toluene-d8, for NMR, 99.8 atom % D
CAS: 2037-26-5 | C7H8 | 100.19 g/mol
Have Questions?
Catalog NumberQuantity
ACR46424010010 mL
Catalog number ACR464240100
Price (MYR)
1,299.00
EA
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
1,299.00
EA
Add to cart
Specifications
Chemical Name or MaterialToluene-d8
Density0.9400 g/mL
Flash Point4°C
Refractive Index1.4940
Specific Gravity0.94