Pyridine-d5, for NMR, 99.5 atom % D
Pyridine-d5, for NMR, 99.5 atom % D
Thermo Scientific Chemicals
Pyridine-d5, for NMR, 99.5 atom % D
CAS: 7291-22-7 | C5H5N | 84.13 g/mol
Have Questions?
Catalog NumberQuantity
ACR46419010010 mL
Catalog number ACR464190100
Price (MYR)
1,531.00
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
1,531.00
Add to cart
Chemical Identifiers
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.7 %
Heavy metals (as Pb)=<0.5 ppm
Chloride (Cl)=<1 ppm
Sulfate (SO4)=<1 ppm