Phosphoric acid, for HPLC, Thermo Scientific Chemicals
Phosphoric acid, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for HPLC, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ACR463861000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS4395-98-6
IUPAC Namepiperidine-4-carbonitrile
Molecular FormulaC6H10N2
InChI KeyFSDNTQSJGHSJBG-UHFFFAOYSA-N
SMILESN#CC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Refractive Index1.4725-1.4785 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow