Duloxetine hydrochloride
Product Image
Thermo Scientific Chemicals
Duloxetine hydrochloride
333.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR463140010
Price (MYR)
616.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
616.00
Add to cart
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Assay>=99.85 % (By difference)
Residue after evaporation=<20 ppm
Hydrogen sulfide (H2S)=<3 ppm
Appearance (Color)Clear colorless