Meropenem trihydrate
Meropenem trihydrate
Thermo Scientific Chemicals
Meropenem trihydrate
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR462810010
Price (MYR)
576.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Specifications
Chemical Name or MaterialMeropenem trihydrate
CAS119478-56-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.